EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Synthesis of YF3: Yb, Er upconverting nanofluorophores using chitosan and their cytotoxicity in MCF-7 cells.
PMID 25453288
Abstract
In this work, we described one pot hydrothermal synthesis of surface modified water soluble YF3: Yb, Er upconverting nanofluorophores (UCNFs) using natural biopolymer chitosan. The obtained nanocrystals have undergone X-ray diffraction (XRD), Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy (SEM), energy dispersive spectroscopy (EDS), high resolution transmission electron microscopy (HRTEM) and photoluminescence (PL) studies. The nanoparticles possess uniform particle size distribution with average size about 27 nm. The cytotoxicity results revealed that the chitosan capped nanoparticles exhibit excellent biocompatibility in human breast cancer cells. In conclusion, the water soluble chitosan capped YF3: Yb, Er nanoparticles could be used as a potential candidate in bio-imaging and therapeutic applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN
40292
Ammonium fluoride solution, semiconductor grade VLSI PURANAL (Honeywell 17600), 40%
H4FN
40208
Ammonium fluoride solution, semiconductor grade PURANAL (Honeywell 17767)
H4FN
263052
Erbium, chips, 99.9% trace rare earth metals basis
Er
693685
Erbium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Er
261084
Erbium, ingot, 99.9% trace rare earth metals basis
Er
263044
Erbium, powder, −40 mesh, 99.9% trace rare earth metals basis
Er
755990
Erbium, dendritic pieces, purified by distillation, 99.95% trace metals basis
Er
GF45743698
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Er
GF15099767
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Er
GF75586494
Erbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Er
GF59778152
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Er
GF14957994
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Er
GF09631043
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Er
GF93107189
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Er
GF34237469
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Er
GF21606386
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Er
GF18382248
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Er
GF33873891
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Er
GF87470253
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Er
GF75856225
Erbium, foil, 25x25mm, thickness 1.0mm, as rolled, 99%
Er
262986
Ytterbium, chips, 99.9% trace rare earth metals basis
Yb
548804
Ytterbium, chunks, 99.9% trace metals basis
Yb
693669
Ytterbium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Yb
261300
Ytterbium, ingot, 99.9% trace rare earth metals basis
Yb
466069
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Yb
GF49006492
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Yb
GF08606516
Ytterbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yb
GF00044682
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yb
GF85959444
Ytterbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yb