EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Multivariate analysis of mainstream tobacco smoke particulate phase by headspace solid-phase micro extraction coupled with comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry.
PMID 25454146
Abstract
A method involving headspace solid-phase microextraction (HS-SPME) and comprehensive two-dimensional gas chromatography (GC×GC) coupled to time-of-flight mass spectrometry (TOFMS) was developed and applied to evaluate profiles of volatile compounds present in mainstream tobacco smoke particulate matter trapped on glass fiber filters. Six SPME fibers were tested for the extraction capacities toward selected compounds, showing the best results for the polyacrylate fiber. The optimization of the extraction conditions was carried out using multivariate response surface methodology. Two cigarette types differing in a filter design were analyzed using optimized conditions. A template was built in order to generate comprehensive chemical information, which conceded obtaining consistent information across 24 chromatograms. Principal component analysis (PCA) allowed a clear differentiation of the studied cigarette types. Fisher ratio analysis allowed identification of compounds responsible for the chemical differences between the cigarette samples. Of the selected 143 most important ones, 134 analytes were reduced by the active carbon filter, while for nine, classical cellulose acetate filter was more efficient.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414565
Divinylbenzene, technical grade, 80%
C10H10
169099
Divinylbenzene, technical grade, 55%
C10H10
C9635
DL-p-Chloroamphetamine hydrochloride, ≥94% (HPLC)
C9H12ClN · HCl
03079
Ethylbenzene, analytical standard
C8H10
PHR1229
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
296848
Ethylbenzene, anhydrous, 99.8%
C8H10
E12508
Ethylbenzene, ReagentPlus®, 99%
C8H10
469300
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
C6H18OSi2
01422
Melting point standard 79-81°C, analytical standard
C10H8
84679
Naphthalene, analytical standard
C10H8
91489
Naphthalene, certified reference material, TraceCERT®
C10H8
184500
Naphthalene, suitable for scintillation, ≥99%
C10H8
147141
Naphthalene, 99%
C10H8
PHR1275
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
C10H8
311421
Perchloric acid, 70%, 99.999% trace metals basis
HClO4
50439
Perchloric acid concentrate, 0.01 M HClO4 in water (0.01N), eluent concentrate for IC
HClO4
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
77613
Phenol, for molecular biology, ~90% (T), liquid
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
PHR1047
Phenol
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
270970
Pyridine, anhydrous, 99.8%
C5H5N
184527
Pyridine, spectrophotometric grade, ≥99%
C5H5N
02486
Pyridine, analytical standard
C5H5N
W296600
Pyridine, ≥99%
C5H5N
24-6510
Pyridine, SAJ first grade, ≥99.0%
C5H5N
PHR1558
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H5N
W347000
Quinoline, ≥97%
C9H7N
241571
Quinoline, reagent grade, 98%
C9H7N