EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Electrochemical biosensor based on glucose oxidase encapsulated within enzymatically synthesized poly(1,10-phenanthroline-5,6-dione).
PMID 25454754
Abstract
This study is focused on the investigation of electrocatalytic effect of glucose oxidase (GOx) immobilized on the graphite rod (GR) electrode. The enzyme modified electrode was prepared by encapsulation of immobilized GOx within enzymatically formed poly(1,10-phenanthroline-5,6-dione) (pPD) film. The electrochemical responses of such enzymatic electrode (pPD/GOx/GR) vs. different glucose concentrations were examined chronoamperometrically in acetate-phosphate buffer solution (A-PBS), pH 6.0, under aerobic or anaerobic conditions. Amperometric signals of the pPD/GOx/GR electrode exhibited well-defined hyperbolic dependence upon glucose concentration. Amperometric signals at 100mM of glucose were 41.17 and 32.27 μA under aerobic and anaerobic conditions, respectively. Amperometric signals of the pPD/GOx/GR electrode decreased by 6% within seven days. The pPD/GOx/GR electrode showed excellent selectivity in the presence of dopamine and uric acid. Furthermore it had a good reproducibility and repeatability with standard deviation of 9.4% and 8.0%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
496383
1,10-Phenanthroline-5,6-dione, 97%
C12H6N2O2