EMAIL THIS PAGE TO A FRIEND
Seizure
Long-term seizure outcome for international consensus classification of hippocampal sclerosis: a survival analysis.
PMID 25455728
Abstract
Surgery is regarded as a common treatment option for patients with mesial temporal lobe epilepsy (MTLE) as a result of hippocampal sclerosis (HS). However, approximately one-third of patients with intractable epilepsy did not become seizure-free after tailored resection strategies. It would be compelling to identify predictive factors of postoperative seizure outcomes. Our aim was to assess the correlation between HS classification and long-term postoperative seizure outcome in patients with MTLE due to HS. To investigate HS classification, semi-quantitative analysis and immunohistochemical staining of neuronal nuclei (NeuN) were performed on 100 postoperative hippocampal specimens. All patients had a 1-7 year postoperative follow-up. The postoperative seizure outcome was evaluated using International League Against Epilepsy (ILAE) outcome classification. Three types of HS were recognized. The highest incidence of initial precipitating injury (IPI) was noted in the HS ILAE type 1 group (53.1%). The most favorable long-term seizure outcome was also noted in the HS ILAE type 1 group. The shortest epilepsy duration was recorded in the HS ILAE type 2 group (mean epilepsy duration=6.64 ± 5.83 years). The completely seizure free rate of patients in all groups declined with an increase in time. Our study for the first time demonstrated a significant correlation between HS ILAE types and long-term postoperative seizure outcome in patients with MTLE due to HS. Therefore, HS ILAE types have predictive value in long-term seizure outcome following epilepsy surgery.