EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Cardiology
Normalization of low-density lipoprotein receptor expression in receptor defective homozygous familial hypercholesterolemia by inhibition of PCSK9 with alirocumab.
PMID 25456764