EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
N-terminal region of myelin basic protein reduces fibrillar amyloid-β deposition in Tg-5xFAD mice.
PMID 25457550
Abstract
Alzheimer's disease is a progressive neurodegenerative disorder that is characterized by extensive deposition of fibrillar amyloid-β (Aβ) in the brain. Previously, myelin basic protein (MBP) was identified to be a potent inhibitor to Aβ fibril formation, and this inhibitory activity was localized to the N-terminal residues 1-64, a fragment designated MBP1. Here, we show that the modest neuronal expression of a fusion protein of the biologically active MBP1 fragment and the enhanced green fluorescent protein (MBP1-EGFP) significantly improved the performance of spatial learning memory in Tg-5xFAD mice, a model of pathologic Aβ accumulation in brain. The levels of insoluble Aβ and fibrillar amyloid were significantly reduced in bigenic Tg-5xFAD/Tg-MBP1-EGFP mice. Quantitative stereological analysis revealed that the reduction in amyloid was because of a reduction in the size of fibrillar plaques rather than a decrease in plaque numbers. The current findings support previous studies showing that MBP1 inhibits Aβ fibril formation inxa0vitro and demonstrate the ability of MBP1 to reduce Aβ pathology and improve behavioral performance.