EMAIL THIS PAGE TO A FRIEND
ACS synthetic biology
High-efficiency multiplex genome editing of Streptomyces species using an engineered CRISPR/Cas system.
PMID 25458909
Abstract
Actinobacteria, particularly those of genus Streptomyces, remain invaluable hosts for the discovery and engineering of natural products and their cognate biosynthetic pathways. However, genetic manipulation of these bacteria is often labor and time intensive. Here, we present an engineered CRISPR/Cas system for rapid multiplex genome editing of Streptomyces strains, demonstrating targeted chromosomal deletions in three different Streptomyces species and of various sizes (ranging from 20 bp to 30 kb) with efficiency ranging from 70 to 100%. The designed pCRISPomyces plasmids are amenable to assembly of spacers and editing templates via Golden Gate assembly and isothermal assembly (or traditional digestion/ligation), respectively, allowing rapid plasmid construction to target any genomic locus of interest. As such, the pCRISPomyces system represents a powerful new tool for genome editing in Streptomyces.