EMAIL THIS PAGE TO A FRIEND
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
A reduced-toxicity regimen is associated with durable engraftment and clinical cure of nonmalignant genetic diseases among children undergoing blood and marrow transplantation with an HLA-matched related donor.
PMID 25459642
Abstract
Blood and marrow transplantation (BMT) is a standard curative therapy for patients with nonmalignant genetic diseases. Myeloablative conditioning has been associated with significant regimen-related toxicity (RRT), whereas reduced-intensity conditioning regimens have been associated with graft failure. In this prospective pilot trial conducted at 2 centers between 2006 and 2013, we report the outcome of 22 patients with nonmalignant genetic diseases who were conditioned with a novel reduced-toxicity regimen: i.v. busulfan (16 mg/kg), alemtuzumab (52 mg/m(2)), fludarabine (140 mg/m(2)), and cyclophosphamide (105 mg/kg). The median age of the study population was 3.5 years (range, 5 months to 26 years). No cases of sinusoidal obstruction syndrome, severe or chronic graft-versus-host disease (GVHD), or primary graft failure were reported. Median time to neutrophil engraftment (>500 cells/μL) and platelet engraftment (>20K cells/μL) were 19 (range, 12 to 50) and 23.5 (range, 14 to 134) days, respectively. The median length of follow-up was 3 years (range, .2 to 6.3). The overall survival rates were 95% at 100 days (95% confidence interval, .72 to .99) and 90% at 6 years (95% confidence interval, .68 to .98). RRT and chronic GVHD are significant barriers to BMT for patients with nonmalignant genetic diseases. This alemtuzumab-based reduced-toxicity regimen appears to be promising with durable engraftment, effective cure of clinical disease, low rates of RRT, and no observed chronic GVHD.
Related Materials