EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Detection, identification and quantification by 1H NMR of adulterants in 150 herbal dietary supplements marketed for improving sexual performance.
PMID 25459948
Abstract
One hundred and fifty dietary supplements (DS) marketed to increase sexual performance were analyzed. All these formulations were claimed to contain only natural compounds, plant extracts and/or vitamins. (1)H NMR spectroscopy was used for detecting the presence of adulterants and for their identification and quantification. Mass spectrometry was used as a complementary method for confirming the chemical structures. 61% of DS were adulterated with phosphodiesterase-5 inhibitors (PDE-5i) (27% with the PDE-5i medicines sildenafil, tadalafil and vardenafil, and 34% with their structurally modified analogues). Among them, 64% contained only one PDE-5i and 36% mixtures of two, three and even four. The amounts of PDE-5i medicines were higher than the maximum recommended dose in 25% of DS tainted with these drugs. Additional 5.5% DS included other drugs for the treatment of sexual dysfunction (yohimbine, flibanserin, phentolamine, dehydroepiandrosterone or testosterone). Some DS (2.5%) contained products (osthole, icariin) extracted from plants known to improve sexual performance. Only 31% of the samples could be considered as true herbal/natural products. A follow-up over time of several DS revealed that manufacturers make changes in the chemical composition of the formulations. Lack of quality or consistent manufacture (contamination possibly due to inadequate cleaning of the manufacturing chain, presence of impurities or degradation products, various compositions of a given DS with the same batch number, inadequate labelling) indicated poor manufacturing practices. In conclusion, this paper demonstrates the power of (1)H NMR spectroscopy as a first-line method for the detection of adulterated herbal/natural DS and the need for more effective quality control of purported herbal DS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7302
4-Acetamidophenol, 98%
C8H9NO2
1003009
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
C8H9NO2
PHR1005
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO2
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
S0625
D-(−)-Salicin, ≥99% (GC)
C13H18O7
79588
D-(−)-Salicin, analytical standard
C13H18O7
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
11317
Methanesulfonic acid, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
CH4O3S
59510
Methanesulfonic acid, for HPLC, ≥99.5% (T)
CH4O3S
64280
Methanesulfonic acid, ≥99.0%, ReagentPlus®
CH4O3S
471356
Methanesulfonic acid, ≥99.0%
CH4O3S
55517
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
CH4O3S
M4141
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
CH4O3S
471348
Methanesulfonic acid solution, 70 wt. % in H2O
CH4O3S
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2
P0300000
Paracetamol, European Pharmacopoeia (EP) Reference Standard
C8H9NO2
Y0000669
Salicin, European Pharmacopoeia (EP) Reference Standard
C13H18O7
S7778
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Na3O4P · 12H2O
71911
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Na3O4P · 12H2O
04277
Sodium phosphate tribasic dodecahydrate, ≥98%
Na3O4P · 12H2O
222003
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Na3O4P · 12H2O