EMAIL THIS PAGE TO A FRIEND
Environmental research
Cadmium exposure and atherosclerotic carotid plaques--results from the Malmö diet and Cancer study.
PMID 25460622
Abstract
Epidemiological studies indicate that cadmium exposure through diet and smoking is associated with increased risk of cardiovascular disease. There are few data on the relationship between cadmium and plaques, the hallmark of underlying atherosclerotic disease. To examine the association between exposure to cadmium and the prevalence and size of atherosclerotic plaques in the carotid artery. A population sample of 4639 Swedish middle-aged women and men was examined in 1991-1994. Carotid plaque was determined by B-mode ultrasound. Cadmium in blood was analyzed by inductively coupled plasma mass spectrometry. Comparing quartile 4 with quartile 1 of blood cadmium, the odds ratio (OR) for prevalence of any plaque was 1.9 (95% confidence interval 1.6-2.2) after adjustment for sex and, age; 1.4 (1.1-1.8) after additional adjustment for smoking status; 1.4 (1.1-1.7) after the addition of education level and life style factors; 1.3 (1.03-1.8) after additional adjustment for risk factors and predictors of cardiovascular disease. No effect modification by sex was found in the cadmium-related prevalence of plaques. Similarly, ORs for the prevalence of small and large plaques were after full adjustment 1.4 (1.0-2.1) and 1.4 (0.9-2.0), respectively. The subgroup of never smokers showed no association between cadmium and atherosclerotic plaques. These results extend previous studies on cadmium exposure and clinical cardiovascular events by adding data on the association between cadmium and underlying atherosclerosis in humans. The role of smoking remains unclear. It may both cause residual confounding and be a source of pro-atherogenic cadmium exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00623
Cadmium, purum p.a., for filling reductors
Cd
265411
Cadmium, foil, thickness 0.5 mm, ≥99.99% trace metals basis
Cd
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF48075453
Cadmium, foil, 195x200mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd