EMAIL THIS PAGE TO A FRIEND
Environmental research
Silver nanoparticles affect on gene expression of inflammatory and neurodegenerative responses in mouse brain neural cells.
PMID 25460644
Abstract
Silver nanoparticles (AgNPs) have antibacterial characteristics, and currently are applied in Ag-containing products. This study found neural cells can uptake 3-5 nm AgNPs, and investigated the potential effects of AgNPs on gene expression of inflammation and neurodegenerative disorder in murine brain ALT astrocytes, microglial BV-2 cells and neuron N2a cells. After AgNPs (5, 10, 12.5 μg/ml) exposure, these neural cells had obviously increased IL-1β secretion, and induced gene expression of C-X-C motif chemokine 13 (CXCL13), macrophage receptor with collagenous structure (MARCO) and glutathione synthetase (GSS) for inflammatory response and oxidative stress neutralization. Additionally, this study found amyloid-β (Aβ) plaques for pathological feature of Alzheimer's disease (AD) deposited in neural cells after AgNPs treatment. After AgNPs exposure, the gene expression of amyloid precursor protein (APP) was induced, and otherwise, neprilysin (NEP) and low-density lipoprotein receptor (LDLR) were reduced in neural cells as well as protein level. These results suggested AgNPs could alter gene and protein expressions of Aβ deposition potentially to induce AD progress in neural cells. It's necessary to take notice of AgNPs distribution in the environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
576832
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265616
Silver, wire, diam. 1.5 mm, 99.9% trace metals basis
Ag