EMAIL THIS PAGE TO A FRIEND
Pharmacological research
EP2 and EP4 receptors mediate PGE2 induced relaxation in murine colonic circular muscle: pharmacological characterization.
PMID 25461458
Abstract
Prostaglandin E2 (PGE2) is a regulator of gastrointestinal motility that might be involved in impaired motor function associated to gut inflammation. The aim of the present work is to pharmacologically characterize responses to exogenous and endogenous PGE2 in the mouse colon targeting EP2 and EP4 receptors. Wild type (WT) and EP2 receptor knockout (EP2-KO) mice were used to characterize PGE2 and butaprost (EP2 receptor agonist) effects on smooth muscle resting membrane potential and myogenic contractility in circularly oriented colonic preparations. In WT animals, PGE2 and butaprost concentration-dependently inhibited spontaneous contractions and hyperpolarized smooth muscle cells. Combination of both EP2 (PF-04418948 0.1μM) and EP4 receptor antagonists (L-161,982 10μM) was needed to block both electrical and mechanical PGE2 responses. Butaprost inhibitory responses (both electrical and mechanical) were totally abolished by PF-04418948 0.1μM. In EP2-KO mice, PGE2 (but not butaprost) concentration-dependently inhibited spontaneous contractions and hyperpolarized smooth muscle cells. In EP2-KO mice, PGE2 inhibition of spontaneous contractility and hyperpolarization was fully antagonized by L-161,982 10μM. In WT animals, EP2 and EP4 receptor antagonists caused a smooth muscle depolarization and an increase in spontaneous mechanical activity. PGE2 responses in murine circular colonic layer are mediated by post-junctional EP2 and EP4 receptors. PF-04418948 and L-161,982 are selective EP2 and EP4 receptor antagonists that inhibit PGE2 responses. These antagonists might be useful pharmacological tools to limit prostaglandin effects associated to dismotility in gut inflammatory processes.