EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Heart failure and cognitive dysfunction.
PMID 25464210
Abstract
It has been estimated that 5.1million Americans suffer from heart failure. Cognitive impairment has been described as a consequence of heart failure in numerous studies spanning the last three decades. This systematic review helps differentiate "cognitive impairment" into mild cognitive impairment, dementia, and delirium. We evaluate the prevalence, pathophysiology, treatment modalities, and possible outcomes previously described with these associations in heart failure. This review also assesses the utility of the different screening modalities and their efficacy as they pertain to recognizing cognitive impairment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4628
DL-Homocysteine, ≥95% (titration)
C4H9NO2S
44925
DL-Homocysteine, analytical standard
C4H9NO2S