EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Remote ischemic preconditioning reduces contrast-induced acute kidney injury in patients with ST-elevation myocardial infarction: a randomized controlled trial.
PMID 25464237
Abstract
Contrast medium-induced acute kidney injury (CI-AKI) is a cardiovascular complication after myocardial infarction treated with emergency percutaneous coronary intervention. The aim of this randomized, sham-controlled trial was to evaluate the impact of remote ischemic preconditioning (RIPC) on CI-AKI in patients with ST-elevation myocardial infarction who received emergency primary percutaneous coronary intervention. Patients with a suspected ST-elevation myocardial infarction were randomly assigned at a 1:1 ratio to receive percutaneous coronary intervention either with (n=63) or without (n=62) RIPC (intermittent arm ischemia through three cycles of 5min of inflation and 5min of deflation of a blood pressure cuff). A total of 47 RIPC patients and 47 control patients met all study criteria. The primary endpoint was the incidence of CI-AKI, which was defined as an increase in serum creatinine >0.5mg/dL or >25% over the baseline value 48-72h after administration of contrast medium. The incidence of CI-AKI was 10% (n=5) in the RIPC group and 36% (n=17) in the control group (p=0.003). The odds ratio of CI-AKI in patients who received RIPC was 0.18 (95% confidence interval: 0.05-0.64; p=0.008). In patients with ST-elevation myocardial infarction, RIPC before percutaneous coronary intervention reduced the incidence of CI-AKI.