EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Aniline-modified porous graphitic carbon for hydrophilic interaction and attenuated reverse phase liquid chromatography.
PMID 25464992
Abstract
Most stationary phases for hydrophilic interaction liquid chromatography (HILIC) and reversed phase liquid chromatography (RPLC) are based on silica. Porous graphitic carbon (PGC) is an attractive alternative to silica-based phases due to its chemical and thermal stability, and unique selectivity. However, native PGC is strongly hydrophobic and in some instances excessively retentive. PGC particles with covalently attached aniline groups (Dimethylaniline-PGC and Aniline-PGC) were synthesized to alter the surface polarity of PGC. First, the diazonium salt of N,N-dimethyl-p-phenylenediamine or 4-nitroaniline was adsorbed onto the PGC surface. The adsorbed salt was reduced with sodium borohydride and (Aniline-PGC only) the nitro group was further reduced with iron powder to the aniline. X-ray photoelectron spectroscopy confirmed the surface functionalities and that these moieties were introduced to the surface at concentrations of 0.9 and 2.1molecules/nm(2), respectively. These modified PGC phases (especially Aniline-PGC) were evaluated as HILIC and reversed phases. The Dimethylaniline-PGC phase displayed only weak HILIC retention of phenolic solutes. In contrast, the Aniline-PGC phase displayed up to nearly a 7-fold increase in HILIC retention vs. an aniline-silica phase and selectivity that differed from 10 other HILIC phases. Introduction of aniline groups to the PGC surface reduced the RPLC retentivity of PGC up to more than 5-fold and improved the separation efficiency up to 6-fold. The chromatographic performance of Aniline-PGC is demonstrated by separations of nucleotides, nucleosides, carboxylic acids, basic pharmaceuticals, and other compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74440
(−)-Nopol, purum, ≥98.0% (sum of enantiomers, GC)
C11H18O
185310
4-Nitroaniline, ≥99%
C6H6N2O2
N2128
4-Nitroaniline, crystalline
C6H6N2O2
31569
4-Nitroaniline, analytical standard
C6H6N2O2
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, eluent additive for LC-MS
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
A9434
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
H4ClN
09718
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
H4ClN
254134
Ammonium chloride, 99.998% trace metals basis
H4ClN
326372
Ammonium chloride, 99.99% trace metals basis
H4ClN
09702
Ammonium chloride, purum p.a., ≥99.0%
H4ClN
PHR1327
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
H4ClN
09724
Ammonium chloride, tested according to Ph.Eur.
H4ClN
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
09683
Ammonium ion solution for ISE, 0.1 M NH4+, analytical standard (for ion-selective electrodes)
H4ClN
09685
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
H4ClN
576794
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
H4Cl14N
242284
Aniline, ACS reagent, ≥99.5%
C6H7N