EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
Placental profiling of UGT1A enzyme expression and activity and interactions with preeclampsia at term.
PMID 25465229
Abstract
Placental UDP-glucuronosyltransferase (UGT) enzymes have critical roles in hormone, nutrient, chemical balance and fetal exposure during pregnancy. Placental UGT1A isoforms were profiled and differences between preeclamptic (PE) and non-PE placental UGT expression determined. In third trimester villous placenta, UGT1A1, 1A4, 1A6 and 1A9 were expressed and active in all specimens (nxa0=xa010), but UGT1A3, 1A5, 1A7, 1A8 and 1A10 were absent. The UGT1A activities were comparable to human liver microsomes per milligram, but placental microsome yields were only 2xa0% of liver (1xa0mg/g of tissue vs. 45xa0mg/g of tissue). For successful PCR, placental collection and processing withinxa060xa0min from delivery, including DNAse and ≥300xa0ng of RNA in reverse transcription were essential and snap freezing in liquid nitrogen immediately was the best preservation method. Although UGT1A6 mRNA was lower in PE (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1381
4-Methylumbelliferone, ≥98%
C10H8O3