EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Cholesterol-based polymeric monolithic columns for capillary liquid chromatography.
PMID 25465372
Abstract
A novel, cholesterol-based polymeric monolithic stationary phase for capillary liquid chromatography, was prepared by thermally initiated in-situ polymerization. Cholesteryl methacrylate (CholMA) was used as a functional monomer and trimethylolpropane trimethacrylate (TRIM) was a cross-linker, while azobisisobutyronitrile (AIBN) was an initiator. Isooctane and toluene were chosen as "poor" and "good" solvent, respectively, as constituents of the porogen solvent. Isocratic elutions of alkylbenzenes and separation of the testing mixture of o-terphenyl and triphenylene were conducted for all of the monoliths to assess their hydrophobicity and planar selectivity characteristic for cholesterol-based stationary phases. The synthesized columns demonstrated efficiency exceeding N=10,000 plates and a plate height of ca. H=30 μm. Column preparation was found to be highly reproducible; the relative standard deviation (RSD) values (n=3) for day-to-day and column-to-column were less than 4.08 and 2.02%, respectively, based on retention factor of alkylbenzenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7313
1-[2-(Trifluoromethyl)phenyl]imidazole
C10H7F3N2
714887
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
C8H12N4
360066
2,2,4-Trimethylpentane, anhydrous, 99.8%
C8H18
650439
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
C8H18
34862
2,2,4-Trimethylpentane, for HPLC, ≥99%
C8H18
360597
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
C8H18
59030
2,2,4-Trimethylpentane, analytical standard
C8H18
59045
2,2,4-Trimethylpentane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C8H18
32291
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
C8H18
15-2200
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
C8H18
15-2190
2,2,4-Trimethylpentane, JIS special grade
C8H18
15-2211
2,2,4-Trimethylpentane, for HPLC
C8H18
258776
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
C8H18
M6514
3-(Trimethoxysilyl)propyl methacrylate, ≥98%
C10H20O5Si
64208
3-(Trimethoxysilyl)propyl methacrylate, Selectophore
C10H20O5Si
440159
3-(Trimethoxysilyl)propyl methacrylate, 98%
C10H20O5Si
650455
Cyclohexane, for HPLC, ≥99.9%
C6H12
34855
Cyclohexane, for HPLC, ≥99.7%
C6H12
33117
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C6H12
179191
Cyclohexane, ACS reagent, ≥99%
C6H12
676861
Cyclohexane, ACS reagent, ≥99%
C6H12
C100307
Cyclohexane, Laboratory Reagent, ≥99.8%
C6H12
44964
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
C8H18
74347
Hydroquinone, certified reference material, TraceCERT®
C6H6O2
1324002
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
C6H6O2
PHR1469
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6O2
64120
Methacryloyl chloride, purum, dist., ≥97.0% (GC), contains ~0.02% 2,6-di-tert-butyl-4-methylphenol as stabilizer
C4H5ClO
523216
Methacryloyl chloride, 97%, contains ~200 ppm monomethyl ether hydroquinone as stabilizer
C4H5ClO
T2800
o-Terphenyl, 99%
C18H14
442721
o-Terphenyl, analytical standard
C18H14