EMAIL THIS PAGE TO A FRIEND
Food chemistry
Photoproducts of carminic acid formed by a composite from Manihot dulcis waste.
PMID 25466082
Abstract
Carbon-TiO2 composites were obtained from carbonised Manihot dulcis waste and TiO2 using glycerol as an additive and thermally treating the composites at 800 °C. Furthermore, carbon was obtained from manihot to study the adsorption, desorption and photocatalysis of carminic acid on these materials. Carminic acid, a natural dye extracted from cochineal insects, is a pollutant produced by the food industry and handicrafts. Its photocatalysis was observed under different atmospheres, and kinetic curves were measured by both UV-Vis and HPLC for comparison, yielding interesting differences. The composite was capable of decomposing approximately 50% of the carminic acid under various conditions. The reaction was monitored by UV-Vis spectroscopy and LC-ESI-(Qq)-TOF-MS-DAD, enabling the identification of some intermediate species. The deleterious compound anthracene-9,10-dione was detected both in N2 and air atmospheres.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229253
Carminic acid
C22H20O13
11298
Carminic acid, analytical standard
C22H20O13