EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of added thiamine on the key odorant compounds and aroma of cooked ham.
PMID 25466091
Abstract
This study shows that thiamine plays a major role in the formation of three key odorants of cooked ham: 2-methyl-3-furanthiol, 2-methyl-3-methyldithiofuran, and bis(2-methyl-3-furyl)disulphide. Analyses revealed that under identical cooking conditions, the productions of these three aroma compounds increase in a closely intercorrelated way when the dose of thiamine increases. Using a specific 2-methyl-3-furanthiol extraction-quantification method, it was possible to relate the amounts of thiamine added in model cooked hams to the amounts of 2-methyl-3-furanthiol produced in the cooking process. Sensory analyses highlighted the role of thiamine as a precursor of cooked ham aroma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
441163
2-Methyl-3-furanthiol, technical grade
C5H6OS