EMAIL THIS PAGE TO A FRIEND
Food chemistry
Physicochemical characterisation of four cherry species (Prunus spp.) grown in China.
PMID 25466099
Abstract
The physicochemical characteristics of four cherry species (Prunus avium, Prunus cerasus, Prunus pseudocerasus and Prunus tomentosa) were evaluated. Inter-species variability was greater than intra-species differences. Glucose and fructose were the main sugars, and malic acid was the main organic acid in all species. Combining HPLC-DAD and LC-ESI-MS/MS technologies, total 25 phenolic components were preliminarily identified. P. avium was characterised by high fruit weight, edible proportion, sugar content and low acid content, which made it suitable for fresh eating. P. cerasus was high in acid content and anthocyanins content, making it a good processing species. P. pseudocerasus had rich flavonols varieties and high proportion of hydrocinnamic acids. P. tomentosa was characterised by high total phenolics content (especially flavonols and tannins) and antioxidant activity, indicating a great developmental potential as a health fruit. The results of the present study might provide theoretical guidance for the further development and utilisation of cherries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1753
(−)-Epicatechin, ≥90% (HPLC)
C15H14O6
68097
(−)-Epicatechin, analytical standard
C15H14O6
E4018
(−)-Epicatechin, ≥98% (HPLC), from green tea
C15H14O6
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
00500590
Chlorogenic acid, primary pharmaceutical reference standard
C16H18O9
Y0000569
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
C16H18O9
C3118
CPNQ, ≥98% (HPLC), solid
C20H17ClN4O3
03940590
Epicatechin, primary pharmaceutical reference standard
C15H14O6
58221
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
ClK
94419
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)
C16H18O9
91213
Neochlorogenic acid, analytical standard
C16H18O9
793590
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
ClK
60126
Potassium chloride, tested according to Ph.Eur.
ClK
PHR1329
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClK
60137 Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl
ClK
60135
Potassium chloride solution, BioUltra, ~3 M in H2O
ClK
60134
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
ClK
60136
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
ClK
60138
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
ClK
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
613053
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
C2H3NaO2