EMAIL THIS PAGE TO A FRIEND
Food chemistry
The tomato sauce making process affects the bioaccessibility and bioavailability of tomato phenolics: a pharmacokinetic study.
PMID 25466100
Abstract
Tomato sauce is the most commonly consumed processed tomato product worldwide, but very little is known about how the manufacturing process may affect the phenolic composition and bioavailability after consumption. In a prospective randomised, cross-over intervention study, we analysed the plasma and urinary levels of tomato phenolic compounds and their metabolites after acute consumption of raw tomatoes and tomato sauce, enriched or not with refined olive oil during production. Respectively, eleven and four phenolic metabolites were found in urine and plasma samples. The plasma concentration and urinary excretion of naringenin glucuronide were both significantly higher after the consumption of tomato sauce than raw tomatoes. The results suggest that the mechanical and thermal treatments during tomato sauce manufacture may help to deliver these potentially bioactive phenolics from the food matrix more effectively than the addition of an oil component, thus increasing their bioavailability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
52186
(±)-Naringenin, analytical standard
C15H12O5
37580
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
C7H6O4
08992
3,4-Dihydroxybenzoic acid, analytical standard
C7H6O4
54630
4-Hydroxybenzoic acid, puriss., ≥99.0% (T)
C7H6O3
240141
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
C7H6O3
PHR1048
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
H20059
4-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
W398608
4-Hydroxybenzoic acid, ≥99%, FG
C7H6O3
C0625
Caffeic acid, ≥98.0% (HPLC)
C9H8O4
60018
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C9H8O4
60020
Caffeic acid, purum, ≥95.0% (HPLC)
C9H8O4
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
00500590
Chlorogenic acid, primary pharmaceutical reference standard
C16H18O9
Y0000569
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
C16H18O9
48640
Ethyl gallate, antioxidant, ≥96.0% (HPLC)
C9H10O5
Y0001013
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
C10H10O4
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
96353
Kaempferol, analytical standard
C15H10O6
00550580
Kaempferol, primary pharmaceutical reference standard
C15H10O6
60010
Kaempferol, ≥97.0% (HPLC)
C15H10O6
K0133
Kaempferol, ≥90% (HPLC), powder
C15H10O6
92649
m-Coumaric acid, analytical standard
C9H8O3
W530098
Naringenin, natural (US), 98%
C15H12O5
C9008
p-Coumaric acid, ≥98.0% (HPLC)
C9H8O3
81377
Poly(tetrafluoroethylene), beads, white
]n
430935
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
]n
430943
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
]n
468096
Poly(tetrafluoroethylene), powder, 35 μm particle size
]n
182478
Poly(tetrafluoroethylene), powder, >40 μm particle size
]n