EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Dibenzazepines and dibenzoxazepines as sodium channel blockers.
PMID 25466191
Abstract
We have identified two related series of dibenzazepine and dibenzoxazepine sodium channel blockers, which showed good potency on Nav1.7 in FLIPR-based and electrophysiological functional assays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
539112
4-Bromo-1-fluoro-2-nitrobenzene, 96%
C6H3BrFNO2