EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Epoxyeicosanoids suppress osteoclastogenesis and prevent ovariectomy-induced bone loss.
PMID 25466887
Abstract
Epoxyeicosatrienoic acids (EETs) are products of arachidonic acid metabolism catalyzed by cytochrome P450 epoxygenases. These small molecules are autocrine and paracrine lipid mediators with important roles in inflammation, cardiovascular function, and angiogenesis. Recent evidence has highlighted EETs as potent promoters of organ regeneration and malignant metastasis. We speculated that EETs might impact osteoclastogenesis and bone loss. Using both in vitro and in vivo studies, we observed that EETs significantly attenuated bone loss and inhibited osteoclast formation and activity, which were associated with a decreased receptor activator of NF-κB ligand (RANKL):osteoprotegerin ratio and serum levels of TNF-α and IL-1β. At the molecular level, EETs abrogated RANKL-induced activation of NF-κB, activator protein-1 (AP-1), and MAPKs, including ERK and JNK, but not p38, during osteoclast formation. EETs also prevented the production of reactive oxygen species (ROS) following RANKL stimulation. As a result, EETs suppressed osteoclast-specific gene expression, including tartrate resistant acid phosphatase (TRAP), cathepsin K (CK), matrix metalloproteinase (MMP)-9, and receptor activator of NF-κB (RANK). In conclusion, our findings demonstrate that EETs inhibit osteoclastogenesis through modulation of multiple pathways both upstream and downstream of RANKL signaling. The administration or stabilized endogenous levels of EETs could represent a novel therapeutic strategy for osteoclast-related disorders, such as rheumatoid arthritis and postmenopausal osteoporosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5641
11,12-Epoxy-(5Z,8Z,14Z)-eicosatrienoic acid, ~100 μg/mL in ethanol, ≥95%
C20H32O3