EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Hydrophilic interaction liquid chromatography method for measuring the composition of aquatic humic substances.
PMID 25467510
Abstract
A hydrophilic interaction liquid chromatography (HILIC) method was developed to measure the composition of humic substances from river, reservoir, and treated wastewater based on their physicochemical properties. The current method fractionates the humic substances into four well-defined groups based on parallel analyses with a neutral and a cationic HILIC column, using mobile phases of varied compositions and pH. The results indicate that: (i) the proportion of carboxylic acids in the humic substances from terrestrial origins is less than half of that from treated wastewater (Jeddah, KSA), (ii) a higher content of basic compounds was observed in the humic substances from treated wastewater and Ribou Reservoir (Cholet, France) than in the sample from Loire River (France), (iii) a higher percentage of hydrophobic macromolecules were found in the humic substances from Loire River than in the other samples, and (iv) humic substances of treated wastewater contained less ionic neutral compounds (i.e., pKa 5-9) than the waters from terrestrial origins. The physicochemical property disparity amongst the compounds in each humic substances sample was also evaluated. The humic substances from the lightly humic Loire river displayed the highest disparity, whereas the highly humic Suwannee river (Georgia, USA) showed the most homogeneous humic substances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
514101
1,2,4-Butanetricarboxylic acid
C7H10O6
135011
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
C6H6O2
442418
4-Methylphenol, analytical standard
C7H8O
242284
Aniline, ACS reagent, ≥99.5%
C6H7N
132934
Aniline, ReagentPlus®, 99%
C6H7N
45326
Aniline, PESTANAL®, analytical standard
C6H7N
51788
Aniline, analytical standard
C6H7N
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
13180
Benzylamine, for GC derivatization, ≥99.0%
C7H9N
407712
Benzylamine, purified by redistillation, ≥99.5%
C7H9N
185701
Benzylamine, ReagentPlus®, 99%
C7H9N
442483
Benzylamine, analytical standard
C7H9N
D0806
Diethylamine, BioXtra
C4H11N
31730
Diethylamine, puriss. p.a., ≥99.5% (GC)
C4H11N
386456
Diethylamine, purified by redistillation, 99.5%
C4H11N