EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Ultra high performance supercritical fluid chromatography coupled with tandem mass spectrometry for screening of doping agents. I: Investigation of mobile phase and MS conditions.
PMID 25467513
Abstract
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes. The best compromise for all compounds in terms of MS sensitivity and chromatographic performance was obtained when adding 2% water and 10mM ammonium formate in the CO2/MeOH mobile phase. Beside mobile phase, the nature of the make-up solvent added for interfacing UHPSFC with MS was also evaluated. Ethanol was found to be the best candidate as it was able to compensate for the negative effect of 2% water addition in ESI- mode and provided a suitable MS response for all doping agents. Sensitivity of the optimized UHPSFC-MS/MS method was finally assessed and compared to the results obtained in conventional UHPLC-MS/MS. Sensitivity was improved by 5-100-fold in UHPSFC-MS/MS vs. UHPLC-MS/MS for 56% of compounds, while only one compound (bumetanide) offered a significantly higher MS response (4-fold) under UHPLC-MS/MS conditions. In the second paper of this series, the optimal conditions for UHPSFC-MS/MS analysis will be employed to screen >100 doping agents in urine matrix and results will be compared to those obtained by conventional UHPLC-MS/MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6011
Acetazolamide, ≥99%, powder
C4H6N4O3S2
97582
Acetazolamide, analytical standard
C4H6N4O3S2
1005004
Acetazolamide, United States Pharmacopeia (USP) Reference Standard
C4H6N4O3S2
A0100000
Acetazolamide, European Pharmacopoeia (EP) Reference Standard
C4H6N4O3S2
Y0001084
Acetazolamide for system suitability, European Pharmacopoeia (EP) Reference Standard
C4H6N4O3S2
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
372331
Ammonium acetate, 99.999% trace metals basis
C2H7NO2
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, eluent additive for LC-MS
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
338818
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
H5NO
320145
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO
09859
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
H5NO
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
B5775
Bendroflumethiazide, analytical standard
C15H14F3N3O4S2
B0400000
Bendroflumethiazide, European Pharmacopoeia (EP) Reference Standard
C15H14F3N3O4S2
B3023
Bumetanide, ≥98%
C17H20N2O5S
B1158000
Bumetanide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O5S
93784
Caffeine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C8H10N4O2