EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
A novel SOD1-ALS mutation separates central and peripheral effects of mutant SOD1 toxicity.
PMID 25468678
Abstract
Transgenic mouse models expressing mutant superoxide dismutase 1 (SOD1) have been critical in furthering our understanding of amyotrophic lateral sclerosis (ALS). However, such models generally overexpress the mutant protein, which may give rise to phenotypes not directly relevant to the disorder. Here, we have analysed a novel mouse model that has a point mutation in the endogenous mouse Sod1 gene; this mutation is identical to a pathological change in human familial ALS (fALS) which results in a D83G change in SOD1 protein. Homozgous Sod1(D83G/D83G) mice develop progressive degeneration of lower (LMN) and upper motor neurons, likely due to the same unknown toxic gain of function as occurs in human fALS cases, but intriguingly LMN cell death appears to stop in early adulthood and the mice do not become paralyzed. The D83 residue coordinates zinc binding, and the D83G mutation results in loss of dismutase activity and SOD1 protein instability. As a result, Sod1(D83G/D83G) mice also phenocopy the distal axonopathy and hepatocellular carcinoma found in Sod1 null mice (Sod1(-/-)). These unique mice allow us to further our understanding of ALS by separating the central motor neuron body degeneration and the peripheral effects from a fALS mutation expressed at endogenous levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1304446
ANTI-BCL11B (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104137
Anti-BCL11B antibody produced in rabbit, affinity isolated antibody
SAB2700225
Anti-BCL11B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA049117
Anti-BCL11B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106486
Anti-BCL11B antibody produced in rabbit, affinity isolated antibody
AV34181
Anti-CSRP3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV34182
Anti-CSRP3 (AB2) antibody produced in rabbit, affinity isolated antibody
GW22036F Anti-CSRP3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500279
Anti-CSRP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA042581
Anti-CSRP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22104F Anti-LMO4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412605
ANTI-LMO4 antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
SAB4503413
Anti-LMO4, C-Terminal antibody produced in rabbit, affinity isolated antibody
PRS4627
Anti-SATB2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4629
Anti-SATB2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001042
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA029543
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0008048M3
Monoclonal Anti-CSRP3 antibody produced in mouse, clone 6D2, purified immunoglobulin, buffered aqueous solution
WH0008543M1
Monoclonal Anti-LMO4 antibody produced in mouse, clone 4H8, purified immunoglobulin, buffered aqueous solution
SAB1404596
Monoclonal Anti-LMO4 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
AMAB90682
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0323, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90680
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0321, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90679
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0320, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90678
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0319, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90635
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0276, purified immunoglobulin, buffered aqueous glycerol solution