PloS one
Ida-1, the Caenorhabditis elegans orthologue of mammalian diabetes autoantigen IA-2, potentially acts as a common modulator between Parkinson's disease and Diabetes: role of Daf-2/Daf-16 insulin like signalling pathway.
PMID 25469508
Abstract
The lack of cure to age associated Parkinson's disease (PD) has been challenging the efforts of researchers as well as health care providers. Recent evidences suggest that diabetic patients tend to show a higher future risk for PD advocating a strong correlation between PD and Diabetes, thus making it intriguing to decipher common genetic cues behind these ailments. We carried out studies on ida-1, the C. elegans orthologue of mammalian type-1 diabetes auto-antigen IA-2 towards achieving its functional workup vis-à-vis various associated endpoints of PD and Diabetes. Employing transgenic C. elegans strain expressing "human" alpha synuclein (NL5901) under normal and increased glucose concentrations, we studied aggregation of alpha synuclein, content of dopamine, expression of dopamine transporter, content of reactive oxygen species, locomotor activity, nuclear translocation of FOXO transcription factor Daf-16, and quantification of Daf2/Daf-16 mRNA. Our findings indicate that ida-1 affords protection in the studied disease conditions as absence of ida-1 resulted in higher alpha-synuclein aggregation under conditions that mimic the blood glucose levels of diabetic patients. We also observed reduced dopamine content, decreased motility, defective Daf-16 translocation and reduced expression of Daf-2 and Daf-16. Our studies establish important function of ida-1 as a modulator in Daf-2/Daf-16 insulin like signalling pathway thus possibly being a common link between PD and Diabetes.