EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[CO intoxication: what to do after the acute episode?].
PMID 25469697
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295116
Carbon monoxide, ≥99.0%
CO