EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Determination of eight artificial sweeteners and common Stevia rebaudiana glycosides in non-alcoholic and alcoholic beverages by reversed-phase liquid chromatography coupled with tandem mass spectrometry.
PMID 25471292
Abstract
The method for the determination of acesulfame-K, saccharine, cyclamate, aspartame, sucralose, alitame, neohesperidin dihydrochalcone, neotame and five common steviol glycosides (rebaudioside A, rebaudioside C, steviol, steviolbioside and stevioside) in soft and alcoholic beverages was developed using high-performance liquid chromatography and tandem mass spectrometry with electrospray ionisation (HPLC-ESI-MS/MS). To the best of our knowledge, this is the first work that presents an HPLC-ESI-MS/MS method which allows for the simultaneous determination of all EU-authorised high-potency sweeteners (thaumatin being the only exception) in one analytical run. The minimalistic sample preparation procedure consisted of only two operations; dilution and centrifugation. Linearity, limits of detection and quantitation, repeatability, and trueness of the method were evaluated. The obtained recoveries at three tested concentration levels varied from 97.0 to 105.7%, with relative standard deviations lower than 4.1%. The proposed method was successfully applied for the determination of sweeteners in 24 samples of different soft and alcoholic drinks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5139
Asp-Phe methyl ester, ≥98%
C14H18N2O5
47135
Aspartame, analytical standard
C14H18N2O5
1043706
Aspartame, United States Pharmacopeia (USP) Reference Standard
C14H18N2O5
A1320000
Aspartame, European Pharmacopoeia (EP) Reference Standard
C14H18N2O5
PHR1381
Aspartame, Pharmaceutical Secondary Standard; Certified Reference Material
C14H18N2O5
41131
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
C7H5NO3S
40927
Neohesperidin, analytical standard
C28H34O15
N1887
Neohesperidin, ≥90%
C28H34O15
30987
Rebaudioside C, analytical standard
C44H70O22
240931
Saccharin, ≥99%
C7H5NO3S
109185
Saccharin, ≥98%
C7H5NO3S
1607007
Saccharin, United States Pharmacopeia (USP) Reference Standard
C7H5NO3S
PHR1341
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
C7H5NO3S
S0040000
Saccharin, European Pharmacopoeia (EP) Reference Standard
C7H5NO3S
19345
Steviol, analytical standard
C20H30O3
69293
Sucralose, ≥98.0% (HPLC)
C12H19Cl3O8
90984
Sucralose, analytical standard
C12H19Cl3O8
1623626
Sucralose, United States Pharmacopeia (USP) Reference Standard
C12H19Cl3O8
PHR1342
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
C12H19Cl3O8
Y0001344
Sucralose, European Pharmacopoeia (EP) Reference Standard
C12H19Cl3O8