EMAIL THIS PAGE TO A FRIEND
Journal of neural transmission (Vienna, Austria : 1996)
5-Hydroxymethylcytosine, the "Sixth Base", during brain development and ageing.
PMID 25471351
Abstract
The epigenome is of fundamental importance for development and ageing. The discovery of 5-hydroxymethylcytosine (5hmC), a further base modification of cytosine beyond 5-methylcytosine, might be of high relevance in understanding the complexity of the human brain, as 5hmC is found in great extent in brain tissue. The aim of this study was to investigate the quantity of 5hmC containing nuclei by immunohistochemistry in human and murine brains at several developmental stages. We performed immunohistochemical stainings on frontal cortex, white matter and cerebellar cortex of 15 healthy controls. Three cases each were assigned to five age groups (foetus, adolescent, adult, elderly, aged). Additionally, cortex and cerebellum of 15 mice sacrificed between day 0 and 120 after birth were investigated. We found marked alterations of 5hmC amount during ageing. In human cortex there was an increase of 5hmC of 50%, in white matter we found an increase of even 200% during ageing. In the cerebellum both internal granular cell layer and molecular cell layer showed a significant increase of 5hmC till adulthood. Purkinje cell nuclei showed constantly positive signals for 5hmC. These data were paralleled in murine brains. Co-labelling of 5hmC and markers for mature and immature cells in murine cerebellar cortex at the age of 7 days revealed that 5hmC was found in mature but not in immature cells. In conclusion, the findings described in this study emphasise the importance of 5hmC in brain development and ageing and will help to better understand the complexity and plasticity of the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300350
Anti-ERBB2 (Ab-1221/1222) antibody produced in rabbit, affinity isolated antibody
SAB4300351
Anti-ERBB2 (Ab-1248) antibody produced in rabbit, affinity isolated antibody
SAB4300349
Anti-ERBB2 (Ab-877) antibody produced in rabbit, affinity isolated antibody
GW22631 Anti-ERBB2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA001383
Anti-ERBB2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500782
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500783
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500787
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500789
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500784
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500785
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500786
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500788
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1303193 ANTI-NEU1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1412730
Anti-NEU1 antibody produced in mouse, purified immunoglobulin, 1 mg/mL
HPA015634
Anti-NEU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV44286
Anti-NEU1 antibody produced in rabbit, IgG fraction of antiserum
HPA021506
Anti-NEU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044204
Anti-NEURL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503563
Anti-NEURL1 antibody produced in rabbit, affinity isolated antibody
E2777 Monoclonal Anti-c-erbB-2 antibody produced in mouse, ~2 mg/mL, clone HER2-96, purified immunoglobulin, buffered aqueous solution
WH0002064M6
Monoclonal Anti-ERBB2 antibody produced in mouse, clone 4B8, purified immunoglobulin, buffered aqueous solution
SAB1403787
Monoclonal Anti-ERBB2 antibody produced in mouse, clone 3B1, purified immunoglobulin, buffered aqueous solution
AMAB90628
Monoclonal Anti-HER2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0269, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90627
Monoclonal Anti-HER2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0268, purified immunoglobulin, buffered aqueous glycerol solution
SAB4100514
Monoclonal Anti-NEU1 antibody produced in mouse, clone NEU1-B591, culture supernatant
SAB1412731
Monoclonal Anti-NEU1 antibody produced in mouse, clone 3F9, purified immunoglobulin, 1 mg/mL