EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Diagnosis and management of community and hospital acquired pneumonia in adults: summary of NICE guidance.
PMID 25471702
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4063 C Reactive Protein from human fluids, buffered aqueous solution