EMAIL THIS PAGE TO A FRIEND
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology
Assay reproducibility and interindividual variation for 15 serum estrogens and estrogen metabolites measured by liquid chromatography-tandem mass spectrometry.
PMID 25472673
Abstract
Interindividual differences in estrogen metabolism may partially account for differences in risks of estrogen-responsive cancers. We conducted a proof-of-performance study to assess the reproducibility of a LC/MS-MS method for measurement of 15 serum estrogens and metabolites (all 15 termed EM) in total (conjugated+unconjugated) and unconjugated forms and describe interindividual variation. Interindividual variation in serum EM profiles was evaluated for 20 premenopausal women, 15 postmenopausal women, and 10 men. Replicate aliquots from 10 premenopausal women, 5 postmenopausal women, and 5 men were assayed eight times over 4 weeks. Components of variance were used to calculate coefficients of variation (CV) and intraclass correlation coefficients (ICC). In postmenopausal women and men, median EM concentrations were similar and substantially lower than that in premenopausal women. Within each sex/menopausal group, the sum of all EM varied 5- to 7-fold across extreme deciles. Some EM had greater variation; total estrone varied approximately 12-fold in premenopausal and postmenopausal women. Unconjugated estradiol varied 17-fold in postmenopausal women but only 5-fold in premenopausal women and men. CVs reflecting variation across replicate measures for individuals were <5% for most EM, but higher in some individuals with a low EM concentration. Overall laboratory CVs for all but one EM were <2% and ICCs were >99% for all EM in each group. The serum EM assay has excellent laboratory reproducibility. In premenopausal women, postmenopausal women, and men, interindividual variation in EM measures is substantially greater than laboratory variation. The serum EM assay is suitable for epidemiologic application. See all the articles in this CEBP Focus section, "Biomarkers, Biospecimens, and New Technologies in Molecular Epidemiology."