EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Cytomegalovirus-mediated activation of pyrimidine biosynthesis drives UDP-sugar synthesis to support viral protein glycosylation.
PMID 25472841
Abstract
Human cytomegalovirus (HCMV) induces numerous changes to the host metabolic network that are critical for high-titer viral replication. We find that HCMV infection substantially induces de novo pyrimidine biosynthetic flux. This activation is important for HCMV replication because inhibition of pyrimidine biosynthetic enzymes substantially decreases the production of infectious virus, which can be rescued through medium supplementation with pyrimidine biosynthetic intermediates. Metabolomic analysis revealed that pyrimidine biosynthetic inhibition considerably reduces the levels of various UDP-sugar metabolites in HCMV-infected, but not mock-infected, cells. Further, UDP-sugar biosynthesis, which provides the sugar substrates required for glycosylation reactions, was found to be induced during HCMV infection. Pyrimidine biosynthetic inhibition also attenuated the glycosylation of the envelope glycoprotein B (gB). Both glycosylation of gB and viral growth were restored by medium supplementation with either UDP-sugar metabolites or pyrimidine precursors. These results indicate that HCMV drives de novo-synthesized pyrimidines to UDP-sugar biosynthesis to support virion protein glycosylation. The importance of this link between pyrimidine biosynthesis and UDP-sugars appears to be partially shared among diverse virus families, because UDP-sugar metabolites rescued the growth attenuation associated with pyrimidine biosynthetic inhibition during influenza A and vesicular stomatitis virus infection, but not murine hepatitis virus infection. In total, our results indicate that viruses can specifically modulate pyrimidine metabolic flux to provide the glycosyl subunits required for protein glycosylation and production of high titers of infectious progeny.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131695
Pyrimidine, ≥98.0%
C4H4N2