EMAIL THIS PAGE TO A FRIEND
Journal of Korean medical science
Gene therapy using hepatocyte growth factor expressing adenovirus improves skin flap survival in a rat model.
PMID 25473214
Abstract
Hepatocyte growth factor (HGF) is a potent angiogenic factor that can stimulate the production of blood vessels in ischemic tissue. We investigated whether gene therapy using HGF-expressing adenovirus could enhance skin flap survival. Sprague-Dawley rats were randomly divided into three groups. Rats were subdermally injected with HGF-expressing adenovirus (HGF virus group), recombinant HGF (rhHGF group), or phosphate buffered saline (PBS group) 2 days before and immediately after 3 × 9 cm caudal flap elevation. The survival area of the skin flap, the ratio of blood flow, CD31-positive vessels and, VEGF expression were examined. Skin flap viability was significantly increased in the HGF virus group compared to the rhHGF and PBS groups (71.4% ± 5.9%, 63.8%± 6.4%, and 39.2% ± 13.0%, respectively) (P = 0.025). Furthermore, the blood flow ratio was significantly increased in the HGF virus group. In the HGF virus group, the number of CD31-positive vessels and vascular endothelial growth factor (VEGF) expression were significantly increased. Gene therapy using HGF-expressing adenovirus increase VEGF expression, the number of viable capillaries, and blood flow to the flap, thereby improving skin flap survival.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
SAB4501868
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501869
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
H1896 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24612.111, purified immunoglobulin, lyophilized powder
H0665 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24516, purified immunoglobulin, lyophilized powder