EMAIL THIS PAGE TO A FRIEND
Neuron
Temporal control of neural progenitors: TGF-β switches the clock forward.
PMID 25475182
Abstract
Little is known about how vertebrate neural progenitors in a given spatial domain change their identity over time. In this issue of Neuron, Dias et al. (2014) discover that hindbrain progenitors switch their output in response to TGF-β signaling.