EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
A correlation between tocopherol content and antioxidant activity in seeds and germinating seeds of soybean cultivars.
PMID 25475360
Abstract
Tocopherols are crucial lipid-soluble antioxidants and essential nutrients. There is increasing interest in the biofortification of crops with vitamin E for reducing micronutrient malnutrition. However, relatively little is known about the development of soybean cultivars with high levels of tocopherol through combined breeding. Tocopherol contents of seeds and germinating seeds of 28 Korean soybean cultivars were analyzed and evaluated for health-promoting activities. Total tocopherol concentrations ranged from 203.9 to 503.1 µg g⁻¹ in seeds and from 20.1 to 230.1 µg g⁻¹ in germinating seeds. The traditional landraces of HaNagari (HN, 503.1 µg g⁻¹), Orialtae (OL, 486.6 µg g⁻¹), SuMoktae (SM, 476.5 µg g⁻¹) and SoRitae (SR, 475.5 µg g⁻¹) showed high levels of tocopherol content. The contents of the four isomers of tocopherol in seeds and germinating seeds were correlated with lipid peroxidation. The γ- and δ-tocopherol contents in seeds were related to 1,1-diphenyl-2-picrylhydrazyl free radical scavenging activity (0.434; P < 0.01 and 0.373; P < 0.05). Total tocopherol content was higher in soybean landraces as compared with modern cultivars developed by cross-breeding. These results suggest that soybean breeding is necessary to increase tocopherol levels.