EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Effects of cytochrome P450 2D6 and 3A5 genotypes and possible coadministered medicines on the metabolic clearance of antidepressant mirtazapine in Japanese patients.
PMID 25475885
Abstract
A personalized treatment approach should be considered with the second-generation psychiatric drug mirtazapine because of high frequencies of side effects, including characteristic drowsiness. Plasma concentrations of mirtazapine in patients are influenced by many factors, including polymorphic cytochrome P450 enzymes contributing to its transformation to 8-hydroxymirtazapine and N-demethylmirtazapine. The aim of this study was to investigate the determinant factors for individual variations of metabolic clearance of mirtazapine using in vitro and in vivo methods. In vitro analyses using liver microsomes from individual humans in correlation assays and recombinantly expressed P450 enzymes revealed that CYP2D6 was the major contributor to mirtazapine 8-hydroxylation with high affinity, and that CYP3A5 catalyzed N-demethylation in a similar high-capacity manner to that of CYP3A4. CYP1A2 was a minor contributor to mirtazapine 8-hydroxylation. Metabolic clearance of mirtazapine determined in substrate depletion assays and mirtazapine 8-hydroxylation activities in individual liver microsomes were significantly lower in CYP2D6 intermediate metabolizers (IM) and poor metabolizers (PM) than in extensive metabolizers (EM) (p<0.05). Trough plasma concentration/dose ratios of mirtazapine from 14 patients were significantly higher in the CYP2D6 IM/PM group than in the EM group and were also higher in the CYP3A5 poor-expressors group than in the expressors group (p<0.05). Mirtazapine clearance in pooled human liver microsomes was inhibited by quinidine (a CYP2D6 inhibitor), ketoconazole (a CYP3A inhibitor), and in combination with risperidone and duloxetine, possible coadministered medicines. These results suggested that mirtazapine metabolic clearance could be variously influenced by the CYP2D6 and CYP3A5 genotypes and coadministered drugs in clinical patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000862
Mianserin for system suitability, European Pharmacopoeia (EP) Reference Standard
C18H20N2 · HCl
M2525
Mianserin hydrochloride
C18H20N2 · HCl
S100
Mianserin hydrochloride, analytical standard, for drug analysis
C18H20N2 · HCl
M1875000
Mianserin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C18H20N2 · HCl
M-919
Mianserin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material
C18H21ClN2
M0443
Mirtazapine, ≥98% (HPLC)
C17H19N3
1444279
Mirtazapine, United States Pharmacopeia (USP) Reference Standard
C17H19N3
Y0000705
Mirtazapine, European Pharmacopoeia (EP) Reference Standard
C17H19N3
Y0000721
Mirtazapine for system suitability, European Pharmacopoeia (EP) Reference Standard
C17H19N3
M-128
Mirtazapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C17H19N3
77440
Phenacetin, ≥98.0% (HPLC)
C10H13NO2
235830
Phenacetin, 97%
C10H13NO2
PHR1249
Phenacetin
C10H13NO2
PHR1094
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13NO2
1514008
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C10H13NO2
R3030
Risperidone, ≥98% (HPLC), powder
C23H27FN4O2
R1000600
Risperidone, European Pharmacopoeia (EP) Reference Standard
C23H27FN4O2
1604654
Risperidone, United States Pharmacopeia (USP) Reference Standard
C23H27FN4O2
Y0000370
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
C23H27FN4O2