EMAIL THIS PAGE TO A FRIEND
Talanta
Quantum-dot submicrobead-based immunochromatographic assay for quantitative and sensitive detection of zearalenone.
PMID 25476288
Abstract
Mycotoxin pollutants are commonly related to cereal products and cause fatal threats in food safety, and therefore require simple and sensitive detection. In this work, quantum-dot (QD) submicrobeads (QBs) were synthesized by encapsulating CdSe/ZnS QDs using the microemulsion technique. The resultant QBs, with approximately 2800 times brighter luminescence than the corresponding QDs, were explored as novel fluorescent probes in the immunochromatographic assay (ICA) for sensitive and quantitative detection of zearalenone (ZEN) in corns. Various parameters that influenced the sensitivity and stability of QB-based ICA (QB-ICA) were investigated and optimized. The optimal QB-ICA exhibits good dynamic linear detection for ZEN over the range of 0.125 ng/mL to 10 ng/mL with a median inhibitory concentration of 1.01±0.09 ng/mL (n=3). The detection limits for ZEN in a standard solution and real corn sample (dilution ratio of 1:30) are 0.0625 ng/mL and 3.6 µg/kg, respectively, which is much better than that of a previously reported gold nanoparticle-based ICA method. Forty-six natural corn samples are assayed using both QB-ICA and enzyme-linked immunosorbent assay. The two methods show a highly significant correlation (R(2)=0.92). Nine ZEN-contaminated samples were further confirmed with liquid chromatography-tandem mass spectrometry (LC-MS/MS), and the QB-ICA results also exhibited good agreement with LC-MS/MS method. In brief, this work demonstrates that QB-ICA is capable of rapid, sensitive screening of toxins in food analysis, and shows great promise for point-of-care testing of other analytes.