EMAIL THIS PAGE TO A FRIEND
Nature communications
Direct cellular delivery of human proteasomes to delay tau aggregation.
PMID 25476420
Abstract
The 26S proteasome is the primary machinery that degrades ubiquitin (Ub)-conjugated proteins, including many proteotoxic proteins implicated in neurodegeneraton. It has been suggested that the elevation of proteasomal activity is tolerable to cells and may be beneficial to prevent the accumulation of protein aggregates. Here we show that purified proteasomes can be directly transported into cells through mesoporous silica nanoparticle-mediated endocytosis. Proteasomes that are loaded onto nanoparticles through non-covalent interactions between polyhistidine tags and nickel ions fully retain their proteolytic activity. Cells treated with exogenous proteasomes are more efficient in degrading overexpressed human tau than endogenous proteasomal substrates, resulting in decreased levels of tau aggregates. Moreover, exogenous proteasome delivery significantly promotes cell survival against proteotoxic stress caused by tau and reactive oxygen species. These data demonstrate that increasing cellular proteasome activity through the direct delivery of purified proteasomes may be an effective strategy for reducing cellular levels of proteotoxic proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7410
Amiloride hydrochloride hydrate, ≥98% (TLC), powder
C6H8ClN7O · HCl · xH2O
C8138
Chlorpromazine hydrochloride, ≥98% (TLC)
C17H19ClN2S · HCl
C0982
Chlorpromazine hydrochloride, meets USP testing specifications
C17H19ClN2S · HCl
C1900000
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C17H19ClN2S · HCl
1125006
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C17H19ClN2S · HCl
31679
Chlorpromazine hydrochloride, VETRANAL, analytical standard
C17H19ClN2S · HCl
46401
Cycloheximide, PESTANAL®, analytical standard
C15H23NO4
18079
Cycloheximide solution, 0.1%, for microbiology
C15H23NO4
G6649
Genistein, synthetic, ≥98% (HPLC), powder
C15H10O5
G6776
Genistein, from Glycine max (soybean), ~98% (HPLC)
C15H10O5
92136
Genistein, analytical standard
C15H10O5
282677
L-Lysine hydrate, 97%
C6H14N2O2 · xH2O
62855
L-Lysine monohydrate, purum, ≥98.0% (NT)
C6H14N2O2 · H2O
L9037
L-Lysine monohydrate, BioReagent, cell culture tested, from non-animal source
C6H14N2O2 · H2O
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
O4511
Okadaic acid from Prorocentrum concavum, ≥90% (HPLC), translucent film
C44H68O13
O9381
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
C44H68O13