EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Novel role for TRPC4 in regulation of macroautophagy by a small molecule in vascular endothelial cells.
PMID 25476892
Abstract
Macroautophagy (autophagy) is an important factor affecting the function of vascular endothelial cells (VECs) and must be tightly regulated in these cells. However, the precise mechanisms underlying this process, particularly in the presence of serum, remain obscure. In this study, we identified trans-3,5,4'-trimethoxystilbene (TMS) as a potent small molecule inducer of autophagy in human umbilical vascular endothelial cells (HUVECs) in the presence of serum. Using high-throughput DNA microarray and siRNA transfection technologies, we demonstrated that TMS induced autophagy by up-regulating the expression of the transient receptor potential canonical channel 4 (TRPC4), an important cation channel in HUVECs. In addition, the overexpression of TRPC4 by plasmid transfection also induced autophagy. Mechanistic studies revealed that the up-regulation of TRPC4 increased the intracellular Ca²⁺ concentration, which, in turn, activated the Ca²⁺/CaMKKβ/AMPK pathway, leading to mTOR inhibition and autophagy. Our study identifies a novel role for TRPC4 in the regulation of autophagy in VECs. TMS is a useful new tool for investigating the molecular mechanism of autophagy in VECs and may serve as a potential lead compound for developing a class of autophagy inducers to treat autophagy-related diseases.