EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Determination of drugs of abuse in bovine dentin using liquid chromatography-electrospray ionization tandem mass spectrometry.
PMID 25476949
Abstract
Drugs deposited in human teeth are well preserved; the spectrum of toxicological investigations may therefore be supplemented by an analysis method for drugs in teeth. A liquid chromatography-electrospray ionization tandem mass spectrometry assay for the detection and quantification of basic drugs of abuse in bovine dentin samples was developed and validated. The drugs and metabolites amphetamine, methamphetamine, methylenedioxymethylamphetamine, methylenedioxyethylamphetamine, codeine, morphine, cocaine and benzoylecgonine were extracted from 50 mg ground dentin powder by ultrasonication for 60 min in methanol 3 times. The extracts were analyzed on a triple-quadrupole mass-spectrometer in multiple reaction monitoring mode. The method was validated and proved to be accurate, precise, selective, specific and stable with good linearity within the calibration range and a lower limit of quantification of 10 to 20 pg/mg. To artificially load bovine dentin samples with drugs, the natural process of de- and remineralization in the oral cavity was mimicked by a pH-cycling experiment. The artificially drug-loaded dentin samples showed drug concentrations of 20 to 80 pg/mg. The method can be applied in further in vitro experiments as well as in post-mortem cases, especially where limited sample tissue is available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5080
Calcium chloride dihydrate, BioXtra, ≥99.0%
CaCl2 · 2H2O
C7902
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
CaCl2 · 2H2O
C2536
Calcium chloride dihydrate, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
CaCl2 · 2H2O
C3306
Calcium chloride dihydrate, for molecular biology, ≥99%
CaCl2 · 2H2O
21097
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
CaCl2 · 2H2O
21098
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
CaCl2 · 2H2O
21102
Calcium chloride dihydrate, tested according to Ph.Eur.
CaCl2 · 2H2O
69785
DL-Lactic acid, ~90% (T)
C3H6O3
02424
Ethanesulfonic acid, purum, ≥99.0% (T)
C2H6O3S
186260
Ethanesulfonic acid, 95%
C2H6O3S
471550
Ethanesulfonic acid solution, 70 wt. % in H2O
C2H6O3S
1356734
Lactic acid, United States Pharmacopeia (USP) Reference Standard
C3H6O3
PHR1215
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O3
T0501
Thymol, ≥98.5%
C10H14O
T1350000
Thymol, European Pharmacopoeia (EP) Reference Standard
C10H14O
PHR1134
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
C10H14O
16254
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
C10H14O
50409
Thymol, Standard for quantitative NMR, TraceCERT®
C10H14O
72477
Thymol, analytical standard
C10H14O
00670590
Thymol, primary pharmaceutical reference standard
C10H14O
89329
Thymol, tested according to Ph.Eur.
C10H14O
W306606
Thymol, FCC, FG
C10H14O