EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
β-Cyclodextrin coated SiO₂@Au@Ag core-shell nanoparticles for SERS detection of PCBs.
PMID 25478906
Abstract
A new type of surface-enhanced Raman scattering (SERS) substrate consisting of β-cyclodextrin (β-CD) coated SiO2@Au@Ag nanoparticles (SiO2@Au@Ag@CD NPs) has been achieved. Our protocol was a simplified approach as the fabrication and modification of the silver shell were realized in a single-step reaction by taking advantage of β-CD as both the reducing and stabilizing agents. The as-synthesized SiO2@Au@Ag@CD NPs were uniform in size and demonstrated high SERS activity and reproducibility. The substrates consisting of the SiO2@Au@Ag@CD NPs were employed for SERS detection of polychlorinated biphenyls (PCBs) including PCB-3, PCB-29 and PCB-77. The SERS detection sensitivity was significantly improved due to enrichment of more PCB molecules captured by β-CD on the substrate surface, as confirmed by the appearance of the new Raman bands which are attributed to the complexes between β-CD and PCBs according to the theoretical simulation. Therefore, this work presents a novel approach to the fabrication of effective SERS substrates that can be employed for rapid determination of trace amounts of PCBs in the environment with high detection sensitivity and recognition selectivity.