EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Effect of thermal denaturation, inhibition, and cleavage of disulfide bonds on the low-frequency Raman and FTIR spectra of chymotrypsin and albumin.
PMID 25478913
Abstract
The analysis of the structure-function relationship is extremely important in the study of proteins. The importance of function-related motions of large parts or subglobules of protein molecules stimulates the spectroscopic study in the low-frequency (terahertz) domain. However, only tentative assignments are available and the spectroscopic data are insufficiently discussed in terms of structural changes. This work is aimed at the analysis of regularities of changes in the low-frequency (100 to 600 cm(-1)) FTIR and Raman spectra of proteins related to their structural modifications. We study the spectra of two proteins with substantially different structures (albumin and chymotrypsin) and the spectra of samples in which the structures of protein molecules are modified using inhibition, thermal denaturation, and cleavage of disulfide bonds. The results indicate that the low-frequency spectral interval can be used to characterize protein conformations. Correlated variations in the intensities of several low-frequency bands are revealed in the spectra of the modified proteins. The strongest spectral changes are caused by thermal denaturation of proteins, and the effect of cleavage of disulfide bonds is generally weaker. It is demonstrated that the inhibitor binding in the active site causes spectral changes that can be compared to the changes induced by thermal denaturation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3142 α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
C4129
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
C7762
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
C6423
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
CHY5S
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
C8946 α-Chymotrypsin from human pancreas, lyophilized powder
C1012 α-Chymotrypsin, Inactivated from bovine pancreas, Type VI (Inactivated with DFP), lyophilized powder, <0.2 units/mg protein
A2514
Albumin from goat, ≥96% (agarose gel electrophoresis)
A2639
Albumin from guinea pig serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3060
Albumin from guinea pig serum, lyophilized powder, essentially fatty acid free
A5409 Albumin from hamster
A3139 Albumin from mouse serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3559 Albumin from mouse serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A1056
Albumin from mouse serum, essentially fatty acid free
A4414 Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A2764 Albumin from porcine serum, lyophilized powder, 96-99% (agarose gel electrophoresis)
A1830
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)
A0764
Albumin from rabbit serum, lyophilized powder, essentially globulin free, ~99% (agarose gel electrophoresis)
A0639
Albumin from rabbit serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A9438 Albumin from rabbit serum, essentially fatty acid-free
A6272
Albumin from rat serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A4538
Albumin from rat serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A6414
Albumin from rat serum, lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)
A2018 Albumin from rat serum, essentially fatty acid free
A3264
Albumin from sheep serum, lyophilized powder
A6289 Albumin from sheep serum, essentially fatty acid free
B2518
Albumin, Acetylated from bovine serum, lyophilized powder, suitable for (for molecular biology)
B8894
Albumin, Acetylated from bovine serum, protease free, for molecular biology
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A2153
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)