EMAIL THIS PAGE TO A FRIEND
Developmental and comparative immunology
A novel crustin from Marsupenaeus japonicus promotes hemocyte phagocytosis.
PMID 25479014
Abstract
Crustins are cationic cysteine-rich antimicrobial peptides (AMPs) that contain multiple domains (glycine-rich, cysteine-rich, or proline-rich) at the N-terminus and whey acidic protein (WAP) domains at the C-terminus. Crustins have multiple functions, including protease inhibition and antimicrobial activity. Other functions of crustins need to be clarified. In this study, a novel crustin with a cysteine-rich region, and a single WAP domain, belonging to type I crustins, was identified in Marsupenaeus japonicus and designated as MjCru I-1. MjCru I-1 was expressed in various tissues. The expression of MjCru I-1 was upregulated in the hemocytes of shrimp challenged with bacteria. MjCru I-1 could bind to bacteria by binding to the cell wall molecules of the bacteria, such as lipopolysaccharide (LPS), peptidoglycan (PGN), and lipoteichoic acid (LTA). The synthesized WAP domain of MjCru I-1 but not synthesized Cys-rich domain has antibacterial and agglutinative activities. Scanning electron microscope assay showed that the bacterial cells treated with sMjCru I-1 appeared to be disrupted and cracked compared with those of the control samples. The knockdown of MjCru I-1 could reduce bacterial clearance and injection of MjCru I-1 could significantly increase the survival rate of shrimp infected with Vibrio anguillarum and Staphylococcus aureus compared with those of the control samples. Further study discovered that MjCru I-1 could increase the hemocyte phagocytosis against V. anguillarum and S. aureus. These results suggest that MjCru I-1 has dual functions, bactericidal and phagocytosis promoting activities, in the antibacterial immunity of shrimp.