PloS one
The eukaryotic elongation factor 1A is critical for genome replication of the paramyxovirus respiratory syncytial virus.
PMID 25479059
Abstract
The eukaryotic translation factor eEF1A assists replication of many RNA viruses by various mechanisms. Here we show that down-regulation of eEF1A restricts the expression of viral genomic RNA and the release of infectious virus, demonstrating a biological requirement for eEF1A in the respiratory syncytial virus (RSV) life cycle. The key proteins in the replicase/transcriptase complex of RSV; the nucleocapsid (N) protein, phosphoprotein (P) and matrix (M) protein, all associate with eEF1A in RSV infected cells, although N is the strongest binding partner. Using individually expressed proteins, N, but not P or M bound to eEF1A. This study demonstrates a novel interaction between eEF1A and the RSV replication complex, through binding to N protein, to facilitate genomic RNA synthesis and virus production.