EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Edaravone prevents lung injury induced by hepatic ischemia-reperfusion.
PMID 25481526
Abstract
Lung injury is a major clinical concern after hepatic ischemia-reperfusion (I/R), due to the production of reactive oxygen species in the reperfused liver. We investigated the efficacy of edaravone, a potent free-radical scavenger, for attenuating lung injury after hepatic I/R. Adult male Sprague-Dawley rats were assigned to shamxa0+xa0normal saline (NS), I/Rxa0+xa0NS, or I/Rxa0+xa0edaravone group. Rats in the I/R groups were subjected to 90xa0min of partial hepatic I/R. Five minutes before reperfusion, 3xa0mg/kg edaravone was administered to the I/Rxa0+xa0edaravone group. After 6xa0h of reperfusion, we evaluated lung histopathology and wet-to-dry ratio. We also measured malondialdehyde (MDA), an indicator of oxidative stress, in the liver and the lung, as well as cytokine messenger RNA expressions in the reperfused liver and plasma cytokine concentrations. Histopathology revealed lung damages after 6xa0h reperfusion of partial ischemic liver. Moreover, a significant increase in lung wet-to-dry ratio was observed. MDA concentration increased in the reperfused liver, but not in the lungs. Edaravone administration attenuated the lung injury and the increase of MDA in the reperfused liver. Edaravone also suppressed the reperfusion-induced increase of interleukin-6 messenger RNA expressions in the liver and plasma interleukin-6 concentrations. Edaravone administration before reperfusion of the ischemic liver attenuates oxidative stress in the reperfused liver and the subsequent lung injury. Edaravone may be beneficial for preventing lung injury induced by hepatic I/R.