EMAIL THIS PAGE TO A FRIEND
Pulmonary pharmacology & therapeutics
Poly (ADP-ribose) polymerase-1: an emerging target in right ventricle dysfunction associated with pulmonary hypertension.
PMID 25481773
Abstract
Recently, inhibition of poly (ADP-ribose) polymerase-1 (PARP1) was shown to be protective in experimental pulmonary hypertension (PH) and prevented right ventricular hypertrophy (RVH) associated with it. However, molecular mechanism behind cardioprotection by PARP1 inhibition in PH still needs detailed exploration. Therefore, effect of inhibition of PARP1 on the right ventricle (RV) dysfunction was studied in monocrotaline (MCT) induced PH model. Following a single dose administration of MCT (60xa0mg/kg, s.c.), male Sprague-Dawley rats were treated with PARP1 inhibitor 1,5-Isoquinolinediol (ISO, 3xa0mg/kg, i.p.) for 35 days for preventive study and from day 21-35 for curative study. RV pressure (RVP) and RVH were measured after 35 days. Histophathological studies, PARP1 activity, mRNA and protein expression were studied in isolated RV. Oxidative and nitosative stress, inflammation and Matrix metalloproteinases (MMPs)/Tissue inhibitor of metalloproteinase 2 (TIMP2) were also assessed. Mitochondrial dysfunction was studied by mitochondrial membrane permeability and estimation of Nicotinamide adenine dinucleotide (NAD) and Adenosine triphosphate (ATP). Apoptosis in RV was assessed by Terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL), caspase 3 activity and cleaved PARP1 expression. PARP1 inhibition significantly reversed the increase in RVP and RVH in both preventive and curative treatment in the MCT-injected rats. ISO lowered oxidative and nitrosative stress and inflammation and restored the balance of MMPs/TIMP2 expression. PARP1 inhibition prevented mitochondrial dysfunction and the release of cell death factors from mitochondria. ISO also decreased apoptosis by decreasing number of TUNEL positive cells, caspase 3 activity and PARP1 cleavage in RV. Thus, PARP1 inhibition ameliorated PH induced RV hypertrophy and may emerge as a new therapeutic target for PH.