EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Interaction of TiO2 nanoparticles with the marine microalga Nitzschia closterium: growth inhibition, oxidative stress and internalization.
PMID 25483108
Abstract
The toxicity of TiO2 engineered nanoparticles (NPs) to the marine microalga Nitzschia closterium was investigated by examining growth inhibition, oxidative stress and uptake. The results indicated that the toxicity of TiO2 particles to algal cells significantly increased with decreasing nominal particle size, which was evidenced by the 96 EC50 values of 88.78, 118.80 and 179.05 mg/L for 21 nm, 60 nm and 400 nm TiO2 particles, respectively. The growth rate was significantly inhibited when the alga was exposed to 5mg/L TiO2 NPs (21 nm). Measurements of antioxidant enzyme activities showed that superoxide dismutase (SOD), catalase (CAT) and peroxidase (POD) activities were first induced and subsequently inhibited following exposure to 5mg/L TiO2 NPs. The depletion of antioxidant enzymes with a concomitant increase in malondialdehyde (MDA) levels and reactive oxygen species (ROS) posed a hazard to membrane integrity. A combination of flow cytometry analysis, transmission electron microscopy and Ti content measurement indicated that TiO2 NPs were internalized in N. closterium cells. The level of extracellular ROS, which was induced by TiO2 NPs under visible light, was negligible when compared with the intracellular ROS level (accounting for less than 6.0% of the total ROS level). These findings suggest that elevated TiO2 nanotoxicity in marine environments is related to increased ROS levels caused by internalization of TiO2 NPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
IRMM531B
Titanium, IRMM® certified Reference Material, 0.5 mm foil
Ti
IRMM531C
Titanium, IRMM® certified Reference Material, 0.5 mm wire
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
348805
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Ti
267481
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
348813
Titanium, foil, thickness 0.1 mm, 99.99% trace metals basis
Ti
348821
Titanium, foil, thickness 0.05 mm, 99.99% trace metals basis
Ti
348848
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
266051
Titanium, rod, diam. 6.35 mm, 99.7% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti
749044
Titanium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Ti
767506
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Ti
GF57414179
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Ti
GF72693883
Titanium, rod, 100mm, diameter 1.5mm, annealed, 99.6+%
Ti