EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Accurately mapping the location of the binding site for the interaction between hepatitis B virus X protein and cytochrome c oxidase III.
PMID 25483779
Abstract
The hepatitis B virus (HBV) X protein (HBx) plays an important pathogenetic role in hepatocarcinoma tumorigenesis. As HBx does not have the ability to bind to double-stranded DNA (dsDNA), protein-protein interaction is crucial for HBx functions. In a previous study, we screened a novel HBx-interacting protein, the cytochrome c oxidase subunit III (COXIII). In the present study, we aimed to accurately map the location of the binding site for the interaction of HBx with COXIII. Two fragments of HBx mutants (X1 aa1-72 and X2 aa1-117) were amplified by polymerase chain reaction (PCR) and separately inserted into the pAS2-1 plasmid. PCR and gene sequencing confirmed the correct insertion of the mutant fragments in the plasmid. The transcription of the mutant fragments in yeast cells was demonstrated by RT-PCR and western blot analysis confirmed that they were accurately translated into fusion proteins. Hybridization on solid medium and the detection of β-galactosidase (β-gal) activity indicated that the binding site for the interaction between HBx and COXIII was located between aa72 and aa117. Specific interactions between the HBxX2 protein and COXIII were verified by co-immunoprecipitation. To the best of our knowledge, this is the first study showing to demonstrate that aa72-117 in HBx is the key region for binding with COXIII.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV50251
Anti-COX3 antibody produced in rabbit, affinity isolated antibody