EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
The spatio-temporal dynamics of PKA activity profile during mitosis and its correlation to chromosome segregation.
PMID 25485503
Abstract
The cyclic adenosine monophosphate dependent kinase protein (PKA) controls a variety of cellular processes including cell cycle regulation. Here, we took advantages of genetically encoded FRET-based biosensors, using an AKAR-derived biosensor to characterize PKA activity during mitosis in living HeLa cells using a single-cell approach. We measured PKA activity changes during mitosis. HeLa cells exhibit a substantial increase during mitosis, which ends with telophase. An AKAREV T>A inactive form of the biosensor and H89 inhibitor were used to ascertain for the specificity of the PKA activity measured. On a spatial point of view, high levels of activity near to chromosomal plate during metaphase and anaphase were detected. By using the PKA inhibitor H89, we assessed the role of PKA in the maintenance of a proper division phenotype. While this treatment in our hands did not impaired cell cycle progression in a drastic manner, inhibition of PKA leads to a dramatic increase in chromososme misalignement on the spindle during metaphase that could result in aneuploidies. Our study emphasizes the insights that can be gained with genetically encoded FRET-based biosensors, which enable to overcome the shortcomings of classical methologies and unveil in vivo PKA spatiotemporal profiles in HeLa cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9501
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
C10H12N5O6P
F6886
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
C22H34O7
F3917
Forskolin, For use in molecular biology applications
C22H34O7
93049
Forskolin, analytical standard
C22H34O7
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
77613
Phenol, for molecular biology, ~90% (T), liquid
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
W322318
Phenol, ≥99%
C6H6O
PHR1047
Phenol
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
P4682
Phenol solution, BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
T1895
Thymidine, powder, BioReagent, suitable for cell culture
C10H14N2O5
T9250
Thymidine, ≥99%
C10H14N2O5
89270
Thymidine, ≥99.0% (HPLC)
C10H14N2O5